Search
Search

COG4| Gene Symbol: COG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592498.2 | XP_005592555.1 | 7 - 8 | 1023 | 101 | |
| XM_005592499.2 | XP_005592556.1 | 7 - 8 | 1226 | 101 | ||
| XM_005592501.2 | XP_005592558.1 | 7 - 8 | 1006 | 101 | ||
| XM_015442845.1 | XP_015298331.1 | 7 - 8 | 954 | 101 | ||
| GenBank mRNA | AB170985.1 | - | 7 - 8 | 1009 | 101 | |
Set Membership: |
|