Search
Search

EPB41L1| Gene Symbol: EPB41L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568892.2 | XP_005568949.1 | 20 - 21 | 4525 | 73 | |
| XM_005568893.2 | XP_005568950.1 | 19 - 20 | 5099 | 73 | ||
| XM_005568894.2 | XP_005568951.1 | 19 - 20 | 4369 | 73 | ||
| XM_005568905.1 | XP_005568962.1 | 16 - 17 | 2224 | 73 | ||
| XM_015458007.1 | XP_015313493.1 | 20 - 21 | 5434 | 73 | ||
| XM_015458008.1 | XP_015313494.1 | 22 - 23 | 2806 | 73 | ||
| XM_015458010.1 | XP_015313496.1 | 21 - 22 | 2770 | 73 | ||
| XM_015458015.1 | XP_015313501.1 | 21 - 22 | 2392 | 73 | ||
| XM_015458016.1 | XP_015313502.1 | 20 - 21 | 2356 | 73 | ||
Set Membership: |
|