Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: STOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005556385.2 | XP_005556442.1 | 3 - 4 | 4839 | 88 | |
XM_005556387.2 | XP_005556444.1 | 5 - 6 | 6878 | 88 | ||
XM_005556389.2 | XP_005556446.1 | 3 - 4 | 6442 | 88 | ||
XM_015451062.1 | XP_015306548.1 | 4 - 5 | 6984 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |