Search
Search

CCDC159| Gene Symbol: CCDC159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588012.2 | XP_005588069.1 | 10 - 11 | 1228 | 59 | |
| XM_005588017.2 | XP_005588074.1 | 8 - 9 | 879 | 59 | ||
| XM_015440535.1 | XP_015296021.1 | 10 - 11 | 1228 | 59 | ||
| XM_015440536.1 | XP_015296022.1 | 10 - 11 | 1235 | 59 | ||
| XM_015440537.1 | XP_015296023.1 | 9 - 10 | 1116 | 59 | ||
| XM_015440538.1 | XP_015296024.1 | 9 - 10 | 997 | 59 | ||
| XM_015440539.1 | XP_015296025.1 | 9 - 10 | 1004 | 59 | ||
| XM_015440540.1 | XP_015296026.1 | 8 - 9 | 879 | 59 | ||
| XM_015440541.1 | XP_015296027.1 | 8 - 9 | 886 | 59 | ||
| XM_015440542.1 | XP_015296028.1 | 8 - 9 | 869 | 59 | ||
Set Membership: |
|