Search
Search

KPNA5| Gene Symbol: KPNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551659.2 | XP_005551716.1 | 8 - 9 | 900 | 107 | |
| XM_005551660.2 | XP_005551717.1 | 7 - 8 | 840 | 107 | ||
| XM_015448586.1 | XP_015304072.1 | 8 - 9 | 949 | 107 | ||
Set Membership: |
|