Search
Search

FARS2| Gene Symbol: FARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554123.2 | XP_005554180.1 | 4 - 5 | 1233 | 88 | |
| XM_015449928.1 | XP_015305414.1 | 4 - 5 | 1232 | 88 | ||
| XM_015449929.1 | XP_015305415.1 | 4 - 5 | 1233 | 88 | ||
| XM_015449930.1 | XP_015305416.1 | 4 - 5 | 1233 | 88 | ||
Set Membership: |
|