Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mf01558429_m1
          See other TRPM3 GE Assays ›
          Gene Symbol:
          TRPM3
          Gene Name:
          transient receptor potential cation channel subfamily M member 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 81285894 - 82195587 on Build M_fas_5.0
          Species:
          Macaca fascicularis
          Species Specific ID (Flybase ID):
          -
          Assay ID Mf01558429_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TRPM3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005581900.2 XP_005581957.1 3 - 4 3848 91
          XM_005581901.2 XP_005581958.1 3 - 4 3848 91
          XM_005581902.2 XP_005581959.1 3 - 4 3848 91
          XM_005581903.2 XP_005581960.1 3 - 4 3848 91
          XM_005581904.2 XP_005581961.1 3 - 4 3848 91
          XM_005581905.2 XP_005581962.1 3 - 4 3848 91
          XM_005581906.2 XP_005581963.1 3 - 4 3848 91
          XM_005581907.2 XP_005581964.1 3 - 4 3848 91
          XM_005581908.2 XP_005581965.1 3 - 4 512 91
          XM_005581909.2 XP_005581966.1 3 - 4 512 91
          XM_005581910.2 XP_005581967.1 3 - 4 512 91
          XM_005581911.2 XP_005581968.1 3 - 4 512 91
          XM_005581912.2 XP_005581969.1 3 - 4 512 91
          XM_005581913.2 XP_005581970.1 3 - 4 512 91
          XM_005581914.2 XP_005581971.2 3 - 4 3286 91
          XM_005581915.2 XP_005581972.1 3 - 4 3846 91
          XM_015437265.1 XP_015292751.1 3 - 4 3287 91
          XM_015437266.1 XP_015292752.1 3 - 4 3287 91
          XM_015437267.1 XP_015292753.1 3 - 4 3287 91
          XM_015437268.1 XP_015292754.1 3 - 4 3286 91
          XM_015437269.1 XP_015292755.1 3 - 4 3286 91
          XM_015437270.1 XP_015292756.1 3 - 4 3285 91
          XM_015437271.1 XP_015292757.1 3 - 4 3286 91
          XM_015437272.1 XP_015292758.1 3 - 4 512 91

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_005581908.2, XM_005581909.2, XM_005581910.2, XM_005581911.2, XM_005581912.2, XM_005581913.2, XM_015437272.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ion channel transporter

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline