Search
Search

RTBDN| Gene Symbol: RTBDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588154.2 | XP_005588211.1 | 6 - 7 | 1291 | 95 | |
| XM_005588156.2 | XP_005588213.1 | 5 - 6 | 766 | 95 | ||
| XM_015440473.1 | XP_015295959.1 | 6 - 7 | 1368 | 95 | ||
| XM_015440474.1 | XP_015295960.1 | 5 - 6 | 841 | 95 | ||
| XM_015440475.1 | XP_015295961.1 | 5 - 6 | 1245 | 95 | ||
| XM_015440476.1 | XP_015295962.1 | 5 - 6 | 1169 | 95 | ||
Set Membership: |
|