Search
Search

NOC3L| Gene Symbol: NOC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565985.2 | XP_005566042.1 | 2 - 3 | 348 | 81 | |
| XM_015456403.1 | XP_015311889.1 | 2 - 3 | 348 | 81 | ||
| XM_015456404.1 | XP_015311890.1 | 2 - 3 | 348 | 81 | ||
Set Membership: |
|