Search
Search

WASF1| Gene Symbol: WASF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551571.2 | XP_005551628.1 | 7 - 8 | 1526 | 133 | |
| XM_005551572.2 | XP_005551629.1 | 7 - 8 | 1499 | 133 | ||
| XM_005551574.2 | XP_005551631.1 | 7 - 8 | 1523 | 133 | ||
| XM_015448558.1 | XP_015304044.1 | 8 - 9 | 1570 | 133 | ||
| GenBank mRNA | AB172114.1 | - | 4 - 5 | 617 | 133 | |
Set Membership: |
|