Search
Search

USP34| Gene Symbol: USP34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575792.2 | XP_005575849.2 | 33 - 34 | 5774 | 96 | |
| XM_015433609.1 | XP_015289095.1 | 33 - 34 | 4696 | 96 | ||
| XM_015433610.1 | XP_015289096.1 | 33 - 34 | 6502 | 96 | ||
| XM_015433611.1 | XP_015289097.1 | 29 - 30 | 4054 | 96 | ||
| XM_015433612.1 | XP_015289098.1 | 33 - 34 | 6580 | 96 | ||
Set Membership: |
|