Search
Search

GYPA| Gene Symbol: GYPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555993.2 | XP_005556050.1 | 4 - 5 | 409 | 74 | |
| XM_005555994.2 | XP_005556051.1 | 5 - 6 | 535 | 74 | ||
| XM_005555995.2 | XP_005556052.1 | 3 - 4 | 352 | 74 | ||
Set Membership: |
|