Search
Search

NARS2| Gene Symbol: NARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579196.2 | XP_005579253.1 | 5 - 6 | 972 | 98 | |
| XM_005579198.2 | XP_005579255.1 | 5 - 6 | 656 | 98 | ||
| XM_005579199.2 | XP_005579256.1 | 5 - 6 | 475 | 98 | ||
| XM_015435539.1 | XP_015291025.1 | 5 - 6 | 967 | 98 | ||
| GenBank mRNA | AB174230.1 | - | 5 - 6 | 632 | 98 | |
Set Membership: |
|