Search
Search

LSM14B| Gene Symbol: LSM14B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569506.2 | XP_005569563.1 | 9 - 10 | 1312 | 108 | |
| XM_005569507.2 | XP_005569564.1 | 8 - 9 | 1239 | 108 | ||
| XM_005569508.2 | XP_005569565.1 | 8 - 9 | 1196 | 108 | ||
| XM_015430034.1 | XP_015285520.1 | 9 - 10 | 1313 | 108 | ||
| XM_015430035.1 | XP_015285521.1 | 9 - 10 | 1278 | 108 | ||
| XM_015430038.1 | XP_015285524.1 | 8 - 9 | 1026 | 108 | ||
Set Membership: |
|