Search
Search

THUMPD3| Gene Symbol: THUMPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547713.2 | XP_005547770.1 | 6 - 7 | 1124 | 134 | |
| XM_005547714.2 | XP_005547771.1 | 6 - 7 | 1454 | 134 | ||
| XM_005547715.2 | XP_005547772.1 | 5 - 6 | 1441 | 134 | ||
| XM_005547716.2 | XP_005547773.1 | 5 - 6 | 1403 | 134 | ||
| XM_015446235.1 | XP_015301721.1 | 5 - 6 | 1552 | 134 | ||
Set Membership: |
|