Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mf02796010_g1
See other LOC102145630 GE Assays ›
Gene Symbol:
LOC102145630
Gene Name:
guanine nucleotide-binding protein G(s) subunit alpha
Gene Aliases:
GNAS
Chromosome Location:
Chr.10: 90730736 - 90802982 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID
Mf02796010_g1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
284.00
Your Price
Online offer:
266.65
266.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: LOC102145630
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_005569451.2
XP_005569508.1
8 - 9
3123
83
XM_005569452.2
XP_005569509.1
7 - 8
3120
83
XM_005569454.2
XP_005569511.1
7 - 8
3078
83
XM_005569455.2
XP_005569512.1
6 - 7
3075
83
XM_005569458.2
XP_005569515.1
8 - 9
545
83
XM_005569459.2
XP_005569516.1
7 - 8
500
83
XM_005569460.2
XP_005569517.1
6 - 7
497
83
XM_005569462.2
XP_005569519.1
7 - 8
464
83
XM_015430001.1
XP_015285487.1
7 - 8
833
83
XM_015430002.1
XP_015285488.1
6 - 7
654
83
XM_015430003.1
XP_015285489.1
7 - 8
1610
83
XM_015430005.1
XP_015285491.1
7 - 8
694
83
XM_015430006.1
XP_015285492.1
7 - 8
613
83
XM_015430007.1
XP_015285493.1
7 - 8
595
83
XM_015430008.1
XP_015285494.1
7 - 8
869
83
XM_015430009.1
XP_015285495.1
7 - 8
703
83
XM_015430010.1
XP_015285496.1
7 - 8
508
83
XM_015430011.1
XP_015285497.1
7 - 8
467
83
XM_015430012.1
XP_015285498.1
7 - 8
827
83
XM_015430013.1
XP_015285499.1
6 - 7
587
83
XM_015430014.1
XP_015285500.1
6 - 7
539
83
XM_015430015.1
XP_015285501.1
6 - 7
824
83
GenBank mRNA
AB220406.1
-
3 - 4
180
83
AB220439.1
-
6 - 7
567
83
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
3' Most
Detect Genomic DNA
Recommended Assay
Inventoried
Probe spans exons
Panther Classification:
Molecular Function -
heterotrimeric G-protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline