Search
Search

PEX11G| Gene Symbol: PEX11G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587753.1 | XP_005587810.1 | 3 - 4 | 453 | 66 | |
| XM_015440601.1 | XP_015296087.1 | 4 - 5 | 1258 | 66 | ||
| XM_015440602.1 | XP_015296088.1 | 3 - 4 | 538 | 66 | ||
| XM_015440603.1 | XP_015296089.1 | 3 - 4 | 592 | 66 | ||
Set Membership: |
|