Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PFDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005557955.2 | XP_005558012.1 | 3 - 4 | 371 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |