Search
Search

SEC24B| Gene Symbol: SEC24B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555656.2 | XP_005555713.1 | 14 - 15 | 2540 | 139 | |
| XM_015450751.1 | XP_015306237.1 | 14 - 15 | 2537 | 139 | ||
| XM_015450753.1 | XP_015306239.1 | 13 - 14 | 2435 | 139 | ||
| XM_015450754.1 | XP_015306240.1 | 13 - 14 | 2432 | 139 | ||
Set Membership: |
|