Search
Search

SAA4| Gene Symbol: SAA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001357502.1 | NP_001344431.1 | 1 - 2 | 82 | 90 | |
| XM_005578464.2 | XP_005578521.1 | 1 - 2 | 112 | 90 | ||
| GenBank mRNA | BB893422.1 | - | 1 - 2 | 139 | 90 | |
Set Membership: |
|