Search
Search

ORC5| Gene Symbol: ORC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550437.1 | XP_005550494.1 | 11 - 12 | 1178 | 116 | |
| XM_015447883.1 | XP_015303369.1 | 11 - 12 | 1191 | 116 | ||
| XM_015447884.1 | XP_015303370.1 | 11 - 12 | 1191 | 116 | ||
| XM_015447885.1 | XP_015303371.1 | 11 - 12 | 1191 | 116 | ||
| XM_015447886.1 | XP_015303372.1 | 11 - 12 | 1191 | 116 | ||
Set Membership: |
|