Search
Search

ASNS| Gene Symbol: ASNS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550143.2 | XP_005550200.1 | 7 - 8 | 1579 | 103 | |
| XM_005550144.2 | XP_005550201.1 | 6 - 7 | 1057 | 103 | ||
| XM_005550145.2 | XP_005550202.1 | 7 - 8 | 1147 | 103 | ||
| XM_005550146.2 | XP_005550203.1 | 6 - 7 | 1543 | 103 | ||
| XM_015447713.1 | XP_015303199.1 | 7 - 8 | 1215 | 103 | ||
| XM_015447714.1 | XP_015303200.1 | 7 - 8 | 1147 | 103 | ||
| GenBank mRNA | AB171262.1 | - | 6 - 7 | 984 | 103 | |
Set Membership: |
|