Search
Search

HDAC5| Gene Symbol: HDAC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584397.2 | XP_005584454.1 | 18 - 19 | 2812 | 65 | |
| XM_005584398.2 | XP_005584455.1 | 18 - 19 | 2814 | 65 | ||
| XM_015438565.1 | XP_015294051.1 | 18 - 19 | 4951 | 65 | ||
| XM_015438566.1 | XP_015294052.1 | 18 - 19 | 4936 | 65 | ||
| XM_015438567.1 | XP_015294053.1 | 18 - 19 | 4924 | 65 | ||
| XM_015438568.1 | XP_015294054.1 | 19 - 20 | 4821 | 65 | ||
| XM_015438569.1 | XP_015294055.1 | 18 - 19 | 2685 | 65 | ||
| XM_015438570.1 | XP_015294056.1 | 18 - 19 | 2679 | 65 | ||
| XM_015438571.1 | XP_015294057.1 | 16 - 17 | 3933 | 65 | ||
| XM_015438572.1 | XP_015294058.1 | 18 - 19 | 2785 | 65 | ||
| XM_015438573.1 | XP_015294059.1 | 18 - 19 | 2787 | 65 | ||
| XM_015438574.1 | XP_015294060.1 | 17 - 18 | 2467 | 65 | ||
| XM_015438575.1 | XP_015294061.1 | 20 - 21 | 3020 | 65 | ||
| XM_015438576.1 | XP_015294062.1 | 19 - 20 | 2970 | 65 | ||
| XM_015438577.1 | XP_015294063.1 | 18 - 19 | 2615 | 65 | ||
| XM_015438578.1 | XP_015294064.1 | 18 - 19 | 4909 | 65 | ||
Set Membership: |
|