Search
Search

GLMN| Gene Symbol: GLMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542737.2 | XP_005542794.1 | 10 - 11 | 1210 | 150 | |
| XM_005542738.2 | XP_005542795.1 | 10 - 11 | 1155 | 150 | ||
Set Membership: |
|