Search
Search

L3MBTL2| Gene Symbol: L3MBTL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567163.2 | XP_005567220.2 | 5 - 6 | 752 | 71 | |
| XM_005567164.2 | XP_005567221.1 | 6 - 7 | 868 | 71 | ||
Set Membership: |
|