Search
Search

VCAN| Gene Symbol: VCAN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557318.2 | XP_005557375.1 | 3 - 4 | 452 | 140 | |
| XM_005557319.2 | XP_005557376.1 | 2 - 3 | 635 | 140 | ||
| XM_005557320.2 | XP_005557377.1 | 2 - 3 | 635 | 140 | ||
Set Membership: |
|