Search
Search

SSBP4| Gene Symbol: SSBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588448.2 | XP_005588505.1 | 15 - 16 | 1316 | 120 | |
| XM_005588452.1 | XP_005588509.1 | 15 - 16 | 1306 | 120 | ||
| XM_005588455.2 | XP_005588512.1 | 14 - 15 | 1279 | 120 | ||
| XM_015440747.1 | XP_015296233.1 | 14 - 15 | 1280 | 120 | ||
| XM_015440748.1 | XP_015296234.1 | 14 - 15 | 1279 | 120 | ||
Set Membership: |
|