Search
Search

LOC101867156| Gene Symbol: LOC101867156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560856.2 | XP_005560913.1 | 4 - 5 | 762 | 117 | |
| XM_015453584.1 | XP_015309070.1 | 4 - 5 | 315 | 117 | ||
Set Membership: |
|