Search
Search

BMPR1A| Gene Symbol: BMPR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565241.2 | XP_005565298.1 | 13 | 2098 | 156 | |
| XM_005565244.2 | XP_005565301.1 | 13 | 2885 | 156 | ||
| XM_015456092.1 | XP_015311578.1 | 15 | 2316 | 156 | ||
| XM_015456093.1 | XP_015311579.1 | 14 | 2132 | 156 | ||
| XM_015456094.1 | XP_015311580.1 | 14 | 2269 | 156 | ||
| XM_015456095.1 | XP_015311581.1 | 13 | 2095 | 156 | ||
| XM_015456096.1 | XP_015311582.1 | 12 | 1970 | 156 | ||
| XM_015456097.1 | XP_015311583.1 | 12 | 1865 | 156 | ||
| XM_015456098.1 | XP_015311584.1 | 12 | 1935 | 156 | ||
Set Membership: |
|