Search
Search

PRR16| Gene Symbol: PRR16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557597.2 | XP_005557654.1 | 2 | 1415 | 99 | |
| XM_005557599.2 | XP_005557656.1 | 3 | 1498 | 99 | ||
| XM_015451814.1 | XP_015307300.1 | 2 | 2140 | 99 | ||
| XM_015451815.1 | XP_015307301.1 | 2 | 1335 | 99 | ||
Set Membership: |
|