Search
Search

CTHRC1| Gene Symbol: CTHRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563854.2 | XP_005563911.1 | 3 - 4 | 758 | 142 | |
| XM_005563855.2 | XP_005563912.1 | 3 - 4 | 891 | 142 | ||
Set Membership: |
|