Search
Search

AGL| Gene Symbol: AGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542615.2 | XP_005542672.1 | 19 - 20 | 2937 | 120 | |
| XM_005542616.2 | XP_005542673.1 | 19 - 20 | 3042 | 120 | ||
| XM_005542617.2 | XP_005542674.1 | 19 - 20 | 2741 | 120 | ||
Set Membership: |
|