Search
Search

MTX3| Gene Symbol: MTX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557262.2 | XP_005557319.1 | 1 - 2 | 96 | 136 | |
| XM_005557264.2 | XP_005557321.1 | 1 - 2 | 96 | 136 | ||
| XM_015451635.1 | XP_015307121.1 | 1 - 2 | 96 | 136 | ||
| XM_015451636.1 | XP_015307122.1 | 1 - 2 | 96 | 136 | ||
Set Membership: |
|