Search
Search

CRMP1| Gene Symbol: CRMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554449.1 | XP_005554506.1 | 5 - 6 | 704 | 115 | |
| XM_005554451.2 | XP_005554508.2 | 5 - 6 | 910 | 115 | ||
| GenBank mRNA | AY650329.1 | - | 4 - 5 | 496 | 115 | |
Set Membership: |
|