Search
Search

P4HA2| Gene Symbol: P4HA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557715.2 | XP_005557772.1 | 6 - 7 | 734 | 115 | |
| XM_005557716.2 | XP_005557773.1 | 5 - 6 | 783 | 115 | ||
| XM_005557717.2 | XP_005557774.1 | 6 - 7 | 734 | 115 | ||
| XM_005557718.2 | XP_005557775.1 | 5 - 6 | 783 | 115 | ||
| XM_015451861.1 | XP_015307347.1 | 5 - 6 | 917 | 115 | ||
| XM_015451862.1 | XP_015307348.1 | 6 - 7 | 643 | 115 | ||
| XM_015451863.1 | XP_015307349.1 | 5 - 6 | 915 | 115 | ||
Set Membership: |
|