Search
Search

MIIP| Gene Symbol: MIIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544762.2 | XP_005544819.1 | 4 - 5 | 705 | 109 | |
| XM_005544764.2 | XP_005544821.1 | 4 - 5 | 705 | 109 | ||
| XM_015442581.1 | XP_015298067.1 | 4 - 5 | 705 | 109 | ||
Set Membership: |
|