Search
Search

GRB7| Gene Symbol: GRB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584038.2 | XP_005584095.1 | 7 - 8 | 1047 | 75 | |
| XM_005584039.2 | XP_005584096.1 | 8 - 9 | 1450 | 75 | ||
| XM_015437902.1 | XP_015293388.1 | 7 - 8 | 1099 | 75 | ||
| XM_015437903.1 | XP_015293389.1 | 7 - 8 | 1100 | 75 | ||
Set Membership: |
|