Search
Search

CSNK1G3| Gene Symbol: CSNK1G3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557632.2 | XP_005557689.1 | 3 - 4 | 1851 | 153 | |
| XM_005557636.2 | XP_005557693.1 | 3 - 4 | 2084 | 153 | ||
| XM_005557637.2 | XP_005557694.1 | 3 - 4 | 2083 | 153 | ||
| XM_005557638.2 | XP_005557695.1 | 3 - 4 | 1851 | 153 | ||
| XM_005557639.2 | XP_005557696.1 | 3 - 4 | 2083 | 153 | ||
| XM_015451822.1 | XP_015307308.1 | 3 - 4 | 2094 | 153 | ||
| XM_015451823.1 | XP_015307309.1 | 3 - 4 | 1940 | 153 | ||
| XM_015451824.1 | XP_015307310.1 | 3 - 4 | 2776 | 153 | ||
| XM_015451825.1 | XP_015307311.1 | 3 - 4 | 2093 | 153 | ||
| XM_015451826.1 | XP_015307312.1 | 3 - 4 | 2093 | 153 | ||
| XM_015451828.1 | XP_015307314.1 | 3 - 4 | 2091 | 153 | ||
| XM_015451829.1 | XP_015307315.1 | 3 - 4 | 2089 | 153 | ||
| XM_015451830.1 | XP_015307316.1 | 3 - 4 | 2082 | 153 | ||
Set Membership: |
|