Search
Search

CDH13| Gene Symbol: CDH13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592649.2 | XP_005592706.1 | 14 - 15 | 2491 | 60 | |
| XM_005592650.2 | XP_005592707.1 | 13 - 14 | 2429 | 60 | ||
| XM_015442730.1 | XP_015298216.1 | 9 - 10 | 1518 | 60 | ||
Set Membership: |
|