Search
Search

COL9A1| Gene Symbol: COL9A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552604.2 | XP_005552661.1 | 15 - 16 | 1488 | 138 | |
| XM_005552605.2 | XP_005552662.1 | 14 - 15 | 1354 | 138 | ||
| XM_005552607.2 | XP_005552664.1 | 10 - 11 | 678 | 138 | ||
| XM_015449246.1 | XP_015304732.1 | 16 - 17 | 1385 | 138 | ||
Set Membership: |
|