Search
Search

COL9A2| Gene Symbol: COL9A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543837.2 | XP_005543894.1 | 3 - 4 | 334 | 62 | |
| XM_015437791.1 | XP_015293277.1 | 2 - 3 | 225 | 62 | ||
Set Membership: |
|