Search
Search

PDE7A| Gene Symbol: PDE7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015454901.1 | XP_015310387.1 | 1 - 2 | 3043 | 151 | |
| XM_015454904.1 | XP_015310390.1 | 2 - 3 | 181 | 151 | ||
| XM_015454905.1 | XP_015310391.1 | 1 - 2 | 3045 | 151 | ||
Set Membership: |
|