Search
Search

NDST2| Gene Symbol: NDST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015456148.1 | XP_015311634.1 | 7 - 8 | 2109 | 123 | |
| XM_015456149.1 | XP_015311635.1 | 7 - 8 | 2108 | 123 | ||
| XM_015456150.1 | XP_015311636.1 | 7 - 8 | 2109 | 123 | ||
| XM_015456151.1 | XP_015311637.1 | 7 - 8 | 2109 | 123 | ||
| XM_015456152.1 | XP_015311638.1 | 7 - 8 | 2109 | 123 | ||
| XM_015456153.1 | XP_015311639.1 | 7 - 8 | 2109 | 123 | ||
| XM_015456154.1 | XP_015311640.1 | 7 - 8 | 2109 | 123 | ||
Set Membership: |
|