Search
Search

CHD1L| Gene Symbol: CHD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542073.2 | XP_005542130.1 | 10 - 11 | 1732 | 104 | |
| XM_005542075.2 | XP_005542132.1 | 7 - 8 | 644 | 104 | ||
Set Membership: |
|