Search
Search

SLC4A11| Gene Symbol: SLC4A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568461.2 | XP_005568518.1 | 12 - 13 | 2089 | 135 | |
| XM_005568462.2 | XP_005568519.1 | 12 - 13 | 2086 | 135 | ||
| XM_005568463.2 | XP_005568520.1 | 12 - 13 | 2083 | 135 | ||
| XM_005568464.2 | XP_005568521.1 | 12 - 13 | 2047 | 135 | ||
| XM_005568465.2 | XP_005568522.1 | 12 - 13 | 2017 | 135 | ||
| XM_005568466.2 | XP_005568523.1 | 12 - 13 | 1860 | 135 | ||
| XM_005568467.2 | XP_005568524.1 | 12 - 13 | 1856 | 135 | ||
| XM_005568469.2 | XP_005568526.1 | 12 - 13 | 1820 | 135 | ||
| XM_015457836.1 | XP_015313322.1 | 11 - 12 | 2394 | 135 | ||
Set Membership: |
|