Search Thermo Fisher Scientific
Gene Symbol: ATOH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015433466.1 | XP_015288952.1 | 2 - 3 | 1266 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |