Search
Search

CORO6| Gene Symbol: CORO6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583329.2 | XP_005583386.1 | 4 - 5 | 742 | 70 | |
| XM_005583330.2 | XP_005583387.1 | 4 - 5 | 681 | 70 | ||
| XM_015438123.1 | XP_015293609.1 | 4 - 5 | 741 | 70 | ||
| XM_015438124.1 | XP_015293610.1 | 4 - 5 | 741 | 70 | ||
| XM_015438125.1 | XP_015293611.1 | 4 - 5 | 681 | 70 | ||
| XM_015438126.1 | XP_015293612.1 | 4 - 5 | 740 | 70 | ||
| XM_015438127.1 | XP_015293613.1 | 3 - 4 | 622 | 70 | ||
Set Membership: |
|