Search
Search

PARD3B| Gene Symbol: PARD3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574020.2 | XP_005574077.1 | 10 - 11 | 1638 | 87 | |
| XM_005574023.2 | XP_005574080.1 | 10 - 11 | 1638 | 87 | ||
| XM_005574025.2 | XP_005574082.1 | 10 - 11 | 1638 | 87 | ||
| XM_005574026.2 | XP_005574083.1 | 7 - 8 | 5632 | 87 | ||
| XM_015432615.1 | XP_015288101.1 | 10 - 11 | 1638 | 87 | ||
| XM_015432617.1 | XP_015288103.1 | 10 - 11 | 1638 | 87 | ||
| XM_015432618.1 | XP_015288104.1 | 10 - 11 | 1638 | 87 | ||
| XM_015432619.1 | XP_015288105.1 | 10 - 11 | 1638 | 87 | ||
Set Membership: |
|