Search
Search

OSBPL1A| Gene Symbol: OSBPL1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587007.2 | XP_005587064.1 | 26 - 27 | 2835 | 110 | |
| XM_005587010.2 | XP_005587067.1 | 25 - 26 | 2564 | 110 | ||
| XM_005587011.2 | XP_005587068.1 | 14 - 15 | 1637 | 110 | ||
| XM_005587012.2 | XP_005587069.1 | 12 - 13 | 1738 | 110 | ||
| XM_015440083.1 | XP_015295569.1 | 24 - 25 | 2473 | 110 | ||
| XM_015440084.1 | XP_015295570.1 | 23 - 24 | 2425 | 110 | ||
| XM_015440085.1 | XP_015295571.1 | 25 - 26 | 2577 | 110 | ||
| XM_015440086.1 | XP_015295572.1 | 24 - 25 | 2520 | 110 | ||
| XM_015440087.1 | XP_015295573.1 | 12 - 13 | 4025 | 110 | ||
| XM_015440088.1 | XP_015295574.1 | 12 - 13 | 1465 | 110 | ||
Set Membership: |
|