Search
Search

SKIL| Gene Symbol: SKIL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546361.2 | XP_005546418.1 | 6 - 7 | 2708 | 101 | |
| XM_005546362.2 | XP_005546419.1 | 6 - 7 | 2570 | 101 | ||
| XM_015445516.1 | XP_015301002.1 | 6 - 7 | 2814 | 101 | ||
| XM_015445517.1 | XP_015301003.1 | 6 - 7 | 2623 | 101 | ||
Set Membership: |
|